N-(5-Amino-2-methylphenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048868-500MG 048868-2.5G
101885-380EA 364.62 USD
101885-380 101885-382
N-(5-Amino-2-methylphenyl)-2-thiophenecarboxamide
N-(5-Amino-2-methylphenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂OS
MDL Number: MFCD09050641
CAS Number: 926195-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR