N-(3-Amino-2-methylphenyl)-2-thiophenecarboxamide

Supplier: Matrix Scientific

048867-500MG 048867-2.5G
101885-376EA 364.62 USD
101885-376 101885-378
N-(3-Amino-2-methylphenyl)-2-thiophenecarboxamide
N-(3-Amino-2-methylphenyl)-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂OS
MDL Number: MFCD07620118
CAS Number: 895979-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR