N-(4-Amino-2-chlorophenyl)isonicotinamide

Supplier: Matrix Scientific

048863-500MG 048863-2.5G
101885-360EA 364.62 USD
101885-360 101885-362
N-(4-Amino-2-chlorophenyl)isonicotinamide
N-(4-Amino-2-chlorophenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MDL Number: MFCD09046144
CAS Number: 926231-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR