N-(4-Amino-2-methoxyphenyl)isonicotinamide

Supplier: Matrix Scientific

048862-500MG 048862-2.5G
101885-356EA 364.62 USD
101885-356 101885-358
N-(4-Amino-2-methoxyphenyl)isonicotinamide
N-(4-Amino-2-methoxyphenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
MDL Number: MFCD02585684
CAS Number: 552814-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR