N-(4-Amino-2-methylphenyl)isonicotinamide

Supplier: Matrix Scientific

048861-500MG 048861-2.5G
101885-352EA 364.62 USD
101885-352 101885-354
N-(4-Amino-2-methylphenyl)isonicotinamide
N-(4-Amino-2-methylphenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O
MDL Number: MFCD09048100
CAS Number: 33974-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR