N-(3-Amino-4-chlorophenyl)isonicotinamide

Supplier: Matrix Scientific

048860-500MG 048860-2.5G
101885-348EA 364.62 USD
101885-348 101885-350
N-(3-Amino-4-chlorophenyl)isonicotinamide
N-(3-Amino-4-chlorophenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MDL Number: MFCD09047392
CAS Number: 926271-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR