N-(5-Amino-2-chlorophenyl)isonicotinamide

Supplier: Matrix Scientific

048859-500MG 048859-2.5G
101885-344EA 364.62 USD
101885-344 101885-346
N-(5-Amino-2-chlorophenyl)isonicotinamide
N-(5-Amino-2-chlorophenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MDL Number: MFCD11568697
CAS Number: 1082186-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR