N-(5-Amino-2-methoxyphenyl)isonicotinamide

Supplier: Matrix Scientific

048858-500MG 048858-2.5G
101885-340EA 364.62 USD
101885-340 101885-342
N-(5-Amino-2-methoxyphenyl)isonicotinamide
N-(5-Amino-2-methoxyphenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
MDL Number: MFCD09048049
CAS Number: 926238-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR