N-(3-Amino-4-fluorophenyl)isonicotinamide

Supplier: Matrix Scientific

048857-500MG 048857-2.5G
101885-336EA 364.62 USD
101885-336 101885-338
N-(3-Amino-4-fluorophenyl)isonicotinamide
N-(3-Amino-4-fluorophenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀FN₃O
MDL Number: MFCD09049599
CAS Number: 926227-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR