N-(5-Amino-2-fluorophenyl)isonicotinamide

Supplier: Matrix Scientific

048856-500MG 048856-2.5G
101885-332EA 364.62 USD
101885-332 101885-334
N-(5-Amino-2-fluorophenyl)isonicotinamide
N-(5-Amino-2-fluorophenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀FN₃O
MDL Number: MFCD09044071
CAS Number: 926227-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR