N-(3-Amino-2-methylphenyl)isonicotinamide

Supplier: Matrix Scientific

048854-500MG 048854-2.5G
101885-324EA 364.62 USD
101885-324 101885-326
N-(3-Amino-2-methylphenyl)isonicotinamide
N-(3-Amino-2-methylphenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O
MDL Number: MFCD09049962
CAS Number: 926258-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR