4'-Amino-2'-chloroacetanilide

Supplier: Matrix Scientific

048839-500MG 048839-2.5G
101885-264EA 364.62 USD
101885-264 101885-266
4'-Amino-2'-chloroacetanilide
4'-Amino-2'-chloroacetanilide

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD00034068
CAS Number: 57556-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR