N-(4-Amino-2-methoxyphenyl)acetamide

Supplier: Matrix Scientific

048838-500MG 048838-2.5G
101885-260EA 364.62 USD
101885-260 101885-262
N-(4-Amino-2-methoxyphenyl)acetamide
N-(4-Amino-2-methoxyphenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD02657492
CAS Number: 5329-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR