N-(5-Amino-2-chlorophenyl)acetamide

Supplier: Matrix Scientific

048837-500MG 048837-2.5G
101885-256EA 364.62 USD
101885-256 101885-258
N-(5-Amino-2-chlorophenyl)acetamide
N-(5-Amino-2-chlorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD08691762
CAS Number: 57946-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR