3'-Amino-4'-fluoroacetanilide

Supplier: Matrix Scientific

048836-500MG 048836-2.5G
101885-252EA 364.62 USD
101885-252 101885-254
3'-Amino-4'-fluoroacetanilide
3'-Amino-4'-fluoroacetanilide

Hazard: Irritant

Formula: C₈H₉FN₂O
MDL Number: MFCD07368688
CAS Number: 113596-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR