3'-Amino-4'-methylacetanilide

Supplier: Matrix Scientific

048835-500MG 048835-2.5G
101885-248EA 384.62 USD
101885-248 101885-250
3'-Amino-4'-methylacetanilide
3'-Amino-4'-methylacetanilide

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD00025202
CAS Number: 6375-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR