1-Chloro-4-(chloromethylsulfonyl)benzene ≥90%

Supplier: Matrix Scientific

Synonyms: p-Chlorophenyl chloromethyl sulphone, (Chloromethyl)(4-chlorophenyl)dioxo-lambda~6~-sulphane

045529-500MG 045529-1G
101871-316EA 346.15 USD
101871-316 101871-318
1-Chloro-4-(chloromethylsulfonyl)benzene ≥90%
(Chloromethyl)(4-chlorophenyl)dioxo-lambda~6~-sulfane

Hazard: Irritant

Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00032558
CAS Number: 5943-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR