6-Hydroxy-1-(4-methylphenyl)-1,3-hexanedione ≥95%
Supplier: Matrix Scientific
045503-500MG
045503-1G
045503-5G
101871-184EA
215.38
USD
101871-184
101871-186
101871-188
6-Hydroxy-1-(4-methylphenyl)-1,3-hexanedione ≥95%
6-Hydroxy-1-(4-methylphenyl)-1,3-hexanedione
Hazard: Irritant
Formula:
C₁₃H₁₆O₃ MW: 220.27 g/mol |
MDL Number:
MFCD06659584 CAS Number: 69745-21-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...