2-Amino-1-morpholino-1-ethanone hydrochloride ≥95%

Supplier: Matrix Scientific

045499-500MG 045499-1G 045499-5G
101871-162EA 69.23 USD
101871-162 101871-164 101871-166
2-Amino-1-morpholino-1-ethanone hydrochloride ≥95%
2-Amino-1-morpholino-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O₂
MDL Number: MFCD07366740
CAS Number: 24152-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR