tert-Butyl-N-(2-morpholino-2-oxoethyl)carbamate ≥95%

Supplier: Matrix Scientific

045496-500MG 045496-1G 045496-5G
101871-144EA 278.46 USD
101871-144 101871-146 101871-148
tert-Butyl-N-(2-morpholino-2-oxoethyl)carbamate ≥95%
tert-Butyl-N-(2-morpholino-2-oxoethyl)carbamate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
MDL Number: MFCD07366739
CAS Number: 114703-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR