3-(tert-Butyl)-2,4-pentanedione, Technical Grade
Supplier: Matrix Scientific
045494-500MG
045494-1G
101871-134EA
315.38
USD
101871-134
101871-136
3-(tert-Butyl)-2,4-pentanedione, Technical Grade
3-(tert-Butyl)-2,4-pentanedione
Hazard: Irritant
Formula:
C₉H₁₆O₂ MW: 156.23 g/mol |
MDL Number:
MFCD01846256 CAS Number: 13221-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...