1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Cyclopropyl-3-(dimethylamino)prop-2-en-1-one

045410-500MG 045410-1G 045410-5G
101870-692EA 161.54 USD
101870-692 101870-694 101870-696
1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one ≥95%
1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one

Hazard: Irritant

Formula: C₈H₁₃NO
MW: 139.2 g/mol
MDL Number: MFCD11840950
CAS Number: 21666-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR