tert-Butyl-2-(4-hydroxyphenoxy)acetate ≥95%

Supplier: Matrix Scientific

045366-500MG 045366-1G
101870-524EA 1109.23 USD
101870-524 101870-526
tert-Butyl-2-(4-hydroxyphenoxy)acetate ≥95%
tert-Butyl-2-(4-hydroxyphenoxy)acetate

Hazard: Irritant

Formula: C₁₂H₁₆O₄
MW: 224.26 g/mol
MDL Number: MFCD12025831
CAS Number: 42806-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR