Methyl-1-(3-fluorobenzyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
045363-500MG
045363-1G
045363-5G
101870-506EA
278.46
USD
101870-506
101870-508
101870-510
Methyl-1-(3-fluorobenzyl)-4-piperidinecarboxylate ≥95%
Methyl-1-(3-fluorobenzyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₈FNO₂ MW: 251.3 g/mol |
MDL Number:
MFCD03425805 CAS Number: 383146-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...