4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-2-(methylsulphanyl)-6-(trifluoromethyl)pyrimidine
045340-500MG
045340-1G
045340-5G
101870-404EA
40
USD
101870-404
101870-406
101870-408
4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine ≥95%
4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine
Hazard: Irritant/Store in Freezer
Formula:
C₆H₄ClF₃N₂S MW: 228.617 g/mol |
MDL Number:
MFCD00269080 CAS Number: 16097-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...