4-Chloro-3-methyl-5-isoxazolamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-4-chloro-3-methylisoxazole

045320-500MG 045320-1G
101870-290EA 90.77 USD
101870-290 101870-292
4-Chloro-3-methyl-5-isoxazolamine ≥95%
4-Chloro-3-methyl-5-isoxazolamine

Hazard: Irritant

Formula: C₄H₅ClN₂O
MW: 132.55 g/mol
MDL Number: MFCD08443950
CAS Number: 166964-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR