N'-Hydroxy-2-[(4-methylphenyl)sulfonyl]ethanimidamide ≥95%

Supplier: Matrix Scientific

045298-500MG 045298-1G
101870-176EA 278.46 USD
101870-176 101870-178
N'-Hydroxy-2-[(4-methylphenyl)sulfonyl]ethanimidamide ≥95%
N'-Hydroxy-2-[(4-methylphenyl)sulfonyl]ethanimidamide

Hazard: Irritant

Formula: C₉H₁₂N₂O₃S
MW: 228.27 g/mol
MDL Number: MFCD00169961
CAS Number: 72660-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR