3-(Diethoxymethyl)-2-ethoxytetrahydro-2H-pyran ≥95%

Supplier: Matrix Scientific

045242-500MG 045242-1G 045242-5G
101869-882EA 278.46 USD
101869-882 101869-884 101869-886
3-(Diethoxymethyl)-2-ethoxytetrahydro-2H-pyran ≥95%
3-(Diethoxymethyl)-2-ethoxytetrahydro-2H-pyran

Hazard: Irritant

Formula: C₁₂H₂₄O₄
MDL Number: MFCD00085003
CAS Number: 69549-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR