4-(tert-Butoxycarbonylamino)-1-butanol ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (4-hydroxybutyl)carbamate, N-Boc-4-aminobutanol, 4-(Boc-amino)-1-butanol, N-(4-Hydroxybutyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(4-Hydroxybutyl)carbamate

045213-1G 045213-5G
101869-718EA 203.08 USD
101869-718 101869-720
4-(tert-Butoxycarbonylamino)-1-butanol ≥95%
4-(tert-Butoxycarbonylamino)-1-butanol

Hazard: Irritant

Formula: C₉H₁₉NO₃
MW: 189.26 g/mol
MDL Number: MFCD01861334
CAS Number: 75178-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR