3-(4-Isopropylphenyl)-2-propen-1-ol ≥95%

Supplier: Matrix Scientific

045198-500MG 045198-1G 045198-5G
101869-646EA 375.38 USD
101869-646 101869-648 101869-650
3-(4-Isopropylphenyl)-2-propen-1-ol ≥95%
3-(4-Isopropylphenyl)-2-propen-1-ol

Hazard: Irritant

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
MDL Number: MFCD08056650
CAS Number: 274907-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR