2-(Phenylmethylene)-1-hydrazinecarboxamide ≥95%

Supplier: Matrix Scientific

045167-500MG 045167-1G 045167-5G
101869-478EA 484.62 USD
101869-478 101869-480 101869-482
2-(Phenylmethylene)-1-hydrazinecarboxamide ≥95%
2-(Phenylmethylene)-1-hydrazinecarboxamide

Hazard: Irritant

Formula: C₈H₉N₃O
MW: 163.18 g/mol
MDL Number: MFCD00025400
CAS Number: 1574-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR