1-(3-Nitro-4-piperidinophenyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

045152-500MG 045152-1G 045152-5G
101869-394EA 278.46 USD
101869-394 101869-396 101869-398
1-(3-Nitro-4-piperidinophenyl)-1-ethanone ≥95%
1-(3-Nitro-4-piperidinophenyl)-1-ethanone

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MW: 248.28 g/mol
MDL Number: MFCD00047468
CAS Number: 30877-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR