Ethyl-1-(4-acetyl-2-nitrophenyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

045151-500MG 045151-1G 045151-5G
101869-388EA 278.46 USD
101869-388 101869-390 101869-392
Ethyl-1-(4-acetyl-2-nitrophenyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(4-acetyl-2-nitrophenyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₅
MDL Number: MFCD00205978
CAS Number: 217489-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR