Methyl-1-(3,4-dichlorobenzyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

045098-500MG 045098-1G 045098-5G
101869-166EA 278.46 USD
101869-166 101869-168 101869-170
Methyl-1-(3,4-dichlorobenzyl)-4-piperidinecarboxylate ≥95%
Methyl-1-(3,4-dichlorobenzyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₇Cl₂NO₂
MDL Number: MFCD03425791
CAS Number: 383146-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR