3-(1-Azepanyl)-1,1,1-trifluoro-2-propanol ≥95%

Supplier: Matrix Scientific

045082-500MG 045082-1G
101869-086EA 921.54 USD
101869-086 101869-088
3-(1-Azepanyl)-1,1,1-trifluoro-2-propanol ≥95%
3-(1-Azepanyl)-1,1,1-trifluoro-2-propanol

Hazard: Irritant

Formula: C₉H₁₆F₃NO
MW: 211.23 g/mol
MDL Number: MFCD02083040
CAS Number: 672952-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR