Methyl-3,4,5-trifluorobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3,4,5-trifluorobenzoate

044939-1G 044939-5G
101868-330EA 21.54 USD
101868-330 101868-332
Methyl-3,4,5-trifluorobenzenecarboxylate ≥95%
Methyl-3,4,5-trifluorobenzenecarboxylate

Hazard: Irritant

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
MDL Number: MFCD06200977
CAS Number: 773873-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR