(1-Benzyl-2-piperidinyl)methanol ≥95%
Supplier: Matrix Scientific
044909-500MG
044909-1G
044909-5G
101868-162EA
181.54
USD
101868-162
101868-164
101868-166
(1-Benzyl-2-piperidinyl)methanol ≥95%
(1-Benzyl-2-piperidinyl)methanol
Hazard: Irritant
Formula:
C₁₃H₁₉NO |
CAS Number:
85387-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...