Ethyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3,4-dihydro-2H-benzo[b][1,4]oxazine-2-carboxylate

044906-500MG 044906-1G 044906-5G
101868-144EA 200 USD
101868-144 101868-146 101868-148
Ethyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%
Ethyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD01663859
CAS Number: 22244-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR