4-Nitro-N-(2-piperidinoethyl)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

044896-500MG 044896-1G 044896-5G
101868-104EA 278.46 USD
101868-104 101868-106 101868-108
4-Nitro-N-(2-piperidinoethyl)benzenecarboxamide ≥95%
4-Nitro-N-(2-piperidinoethyl)benzenecarboxamide

Hazard: Irritant

Formula: C₁₄H₁₉N₃O₃
MDL Number: MFCD00503226
CAS Number: 1664-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR