Ethyl-1-(4-methoxybenzoyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
044877-500MG
044877-1G
044877-5G
101868-006EA
181.54
USD
101868-006
101868-008
101868-010
Ethyl-1-(4-methoxybenzoyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(4-methoxybenzoyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁NO₄ |
MDL Number:
MFCD01213113 CAS Number: 349397-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...