Ethyl-1-(4-chlorobenzoyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044876-500MG 044876-1G 044876-5G
101868-000EA 181.54 USD
101868-000 101868-002 101868-004
Ethyl-1-(4-chlorobenzoyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(4-chlorobenzoyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₈ClNO₃
MW: 295.77 g/mol
MDL Number: MFCD01213111
CAS Number: 349397-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR