2-Chloro-5-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1,3-thiazole ≥95%
Supplier: Matrix Scientific
044862-500MG
044862-1G
044862-5G
101867-934EA
375.38
USD
101867-934
101867-936
101867-938
2-Chloro-5-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1,3-thiazole ≥95%
2-Chloro-5-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1,3-thiazole
Hazard: Irritant
Formula:
C₉H₁₀ClN₃S |
MDL Number:
MFCD02186589 CAS Number: 477713-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...