2-Chloro-6-cyclopropyl-4-(trifluoromethyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

044835-500MG 044835-1G
101867-814EA 556.92 USD
101867-814 101867-816
2-Chloro-6-cyclopropyl-4-(trifluoromethyl)nicotinonitrile ≥95%
2-Chloro-6-cyclopropyl-4-(trifluoromethyl)nicotinonitrile

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂
MDL Number: MFCD01316492
CAS Number: 478049-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR