2-(Allyloxy)-5-chlorobenzaldehyde ≥95%

Supplier: Matrix Scientific

044787-500MG 044787-1G 044787-5G
101867-592EA 375.38 USD
101867-592 101867-594 101867-596
2-(Allyloxy)-5-chlorobenzaldehyde ≥95%
2-(Allyloxy)-5-chlorobenzaldehyde

Hazard: Irritant

Formula: C₁₀H₉ClO₂
CAS Number: 152842-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR