N-Allyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine ≥95%

Supplier: Matrix Scientific

044780-500MG 044780-1G 044780-5G
101867-564EA 253.85 USD
101867-564 101867-566 101867-568
N-Allyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine ≥95%
N-Allyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₉H₈ClF₃N₂
MW: 236.63 g/mol
MDL Number: MFCD00664527
CAS Number: 338761-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR