1-(4-Nitrophenyl)-4-piperidinol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Nitrophenyl)piperidin-4-ol, 4-Hydroxy-1-(4-nitrophenyl)piperidine

044764-500MG 044764-1G 044764-5G
101867-484EA 47.69 USD
101867-484 101867-486 101867-488
1-(4-Nitrophenyl)-4-piperidinol ≥95%
1-(4-Nitrophenyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MDL Number: MFCD00215518
CAS Number: 79421-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR