N'-(6-Benzyl-3-cyano-4,5,6,7-tetrahydrothieno[2,3-c]pyridin-2-yl)-N,N-dimethyliminoformamide ≥95%

Supplier: Matrix Scientific

044684-500MG 044684-1G
101867-088EA 375.38 USD
101867-088 101867-090
N'-(6-Benzyl-3-cyano-4,5,6,7-tetrahydrothieno[2,3-c]pyridin-2-yl)-N,N-dimethyliminoformamide ≥95%
N'-(6-Benzyl-3-cyano-4,5,6,7-tetrahydrothieno[2,3-c]pyridin-2-yl)-N,N-dimethyliminoformamide

Hazard: Irritant

Formula: C₁₈H₂₀N₄S
MW: 324.45 g/mol
MDL Number: MFCD11553021
CAS Number: 1092352-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR