2-Iodophenyl acetate ≥95%

Supplier: Matrix Scientific

044682-500MG 044682-1G 044682-5G
101867-076EA 118.46 USD
101867-076 101867-078 101867-080
2-Iodophenyl acetate ≥95%
2-Iodophenyl acetate

Hazard: Irritant

Formula: C₈H₇IO₂
MDL Number: MFCD07780625
CAS Number: 32865-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR