3-Nitro-4-(1-piperidinyl)benzaldehyde ≥95%
Supplier: Matrix Scientific
044666-500MG
044666-1G
044666-5G
101867-004EA
278.46
USD
101867-004
101867-006
101867-008
3-Nitro-4-(1-piperidinyl)benzaldehyde ≥95%
3-Nitro-4-(1-piperidinyl)benzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₃ MW: 234.26 g/mol |
MDL Number:
MFCD00831780 CAS Number: 39911-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...