Diethyl-2-cyclohexylmalonate ≥95%

Supplier: Matrix Scientific

044644-500MG 044644-1G 044644-5G
101866-876EA 338.46 USD
101866-876 101866-878 101866-880
Diethyl-2-cyclohexylmalonate ≥95%
Diethyl-2-cyclohexylmalonate

Hazard: Irritant

Formula: C₁₃H₂₂O₄
MDL Number: MFCD00019363
CAS Number: 2163-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR